Gene Transcripts

Gene Symbol
ACSL3
Entrez Gene ID
2181
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
ACS3, FACL3, PRO2194
Chromosome Location
Chr. 2: 222860984 - 222944624 on Build GRCh38
UniGene ID
Hs.655772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004457.3 NP_004448.2 16 2349
NM_203372.1 NP_976251.1 15 2242
XM_017003584.1 XP_016859073.1 14 2161
GenBank mRNA AB061712.1 - 16 2317
AF116690.1 - 4 438
AK001471.1 - 11 1268
AK023191.1 - 13 1894
AK314621.1 - 16 2327
BC032144.2 - 3 230
BC041692.1 - 16 2341 5226905
D89053.1 - 14 2040

View Full Product Details