Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000487.5 | NP_000478.3 | 8 | 2448 | |
NM_001085425.2 | NP_001078894.2 | 9 | 2343 | ||
NM_001085426.2 | NP_001078895.2 | 9 | 2295 | ||
NM_001085427.2 | NP_001078896.2 | 9 | 2250 | ||
NM_001085428.2 | NP_001078897.1 | 8 | 2058 |