Gene Transcripts
- Gene Symbol
- ANKRD62P1-PARP4P3
- Entrez Gene ID
- 23783
- Gene Name
- ANKRD62P1-PARP4P3 readthrough, transcribed pseudogene
- Gene Aliases
- VWFP1-ANKRD62P1-PARP4P3
- Chromosome Location
- Chr. 22: 16653708 - 16675540 on Build GRCh38
- UniGene ID
- Hs.627452
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040115.1 |
|
1 |
183 |
|
GenBank mRNA |
DN918251.1 |
- |
1 |
95 |
|
- Gene Symbol
- ANKRD62
- Entrez Gene ID
- 342850
- Gene Name
- ankyrin repeat domain 62
- Gene Aliases
- DKFZp779B1634
- Chromosome Location
- Chr. 18: 12093796 - 12181684 on Build GRCh38
- UniGene ID
- Hs.448906
- Species
- Homo sapiens
View Full Product Details