Gene Transcripts

Gene Symbol
C6orf52
Entrez Gene ID
347744
Gene Name
chromosome 6 open reading frame 52
Gene Aliases
Chromosome Location
Chr. 6: 10671418 - 10694797 on Build GRCh38
UniGene ID
Hs.61389
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145020.1 NP_001138492.1 3 342
NR_026736.1 2 242
NR_026737.1 2 260
XM_006715088.3 XP_006715151.1 3 511
XM_011514572.2 XP_011512874.1 2 307
XM_011514574.2 XP_011512876.1 2 307
XM_011514575.2 XP_011512877.1 3 493
GenBank mRNA BC016820.1 - 2 156
BX280692.1 - 2 242

View Full Product Details