Gene Transcripts

Gene Symbol
RGS9BP
Entrez Gene ID
388531
Gene Name
regulator of G-protein signaling 9 binding protein
Gene Aliases
PERRS, R9AP, RGS9
Chromosome Location
Chr. 19: 32675407 - 32678300 on Build GRCh38
UniGene ID
Hs.528491
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207391.2 NP_997274.2 1 1341
GenBank mRNA AK124499.1 - 1 1341
AK127646.1 - 1 898

View Full Product Details