Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207644.2 | NP_997527.2 | 4 | 1034 | |
GenBank mRNA | AK097054.1 | - | 4 | 2750 | |
BC017595.1 | - | 3 | 1282 | ||
BC032388.1 | - | 4 | 995 | ||
BC108684.1 | - | 4 | 1036 | 6471196 | |
DB535827.2 | - | 1 | 524 |