Gene Transcripts

Gene Symbol
OR7A10
Entrez Gene ID
390892
Gene Name
olfactory receptor family 7 subfamily A member 10
Gene Aliases
BC85395_3, OR19-18
Chromosome Location
Chr. 19: 14840948 - 14841877 on Build GRCh38
UniGene ID
Hs.553776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005190.1 NP_001005190.1 1 229

View Full Product Details