Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032898.4 | NP_116287.2 | 2 | 553 | |
XM_005269370.4 | XP_005269427.1 | 2 | 528 | ||
XM_011513246.2 | XP_011511548.1 | 2 | 289 | ||
GenBank mRNA | AK058155.1 | - | 2 | 456 | |
AK314070.1 | - | 2 | 522 | ||
AY099509.1 | - | 2 | 483 | ||
BC007827.2 | - | 2 | 482 | 3533603 |