Gene Transcripts

Gene Symbol
C17orf97
Entrez Gene ID
400566
Gene Name
chromosome 17 open reading frame 97
Gene Aliases
CK20, LIAT1
Chromosome Location
Chr. 17: 410327 - 414666 on Build GRCh38
UniGene ID
Hs.499607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013672.4 NP_001013694.4 2 601
GenBank mRNA AK128660.1 - 601
BC057385.1 - 2 552
BC070116.1 - 2 527
BC141806.1 - 2 600 40147362
KR780251.1 - 2 601

View Full Product Details