Gene Transcripts

Gene Symbol
CBL
Entrez Gene ID
867
Gene Name
Cbl proto-oncogene
Gene Aliases
C-CBL, CBL2, FRA11B, NSLL, RNF55
Chromosome Location
Chr. 11: 119206276 - 119308149 on Build GRCh38
UniGene ID
Hs.504096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005188.3 NP_005179.2 9 1441
XM_011543057.2 XP_011541359.1 9 1441
XM_017018511.1 XP_016874000.1 8 876
GenBank mRNA BC132733.1 - 9 1370
BC136463.1 - 9 1370 9020452
X57110.1 - 9 1447

View Full Product Details