Gene Transcripts

Gene Symbol
ARL11
Entrez Gene ID
115761
Gene Name
ADP ribosylation factor like GTPase 11
Gene Aliases
ARLTS1
Chromosome Location
Chr. 13: 49628299 - 49633872 on Build GRCh38
UniGene ID
Hs.558599
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138450.5 NP_612459.1 2 728
XM_005266253.3 XP_005266310.1 3 637
GenBank mRNA AF441378.1 - 2 728
AK091249.1 - 2 539
BC013150.1 - 2 507 4340470

View Full Product Details