Gene Transcripts

Gene Symbol
MS4A13
Entrez Gene ID
503497
Gene Name
membrane spanning 4-domains A13
Gene Aliases
Chromosome Location
Chr. 11: 60515382 - 60543424 on Build GRCh38
UniGene ID
Hs.523704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012417.2 NP_001012417.2 4 499
NM_001100909.1 NP_001094379.1 4 499
GenBank mRNA AY324188.1 - 4 499
AY324190.1 - 4 499
BC140799.1 - 4 373 9021667

View Full Product Details