Gene Transcripts

Gene Symbol
ARRDC5
Entrez Gene ID
645432
Gene Name
arrestin domain containing 5
Gene Aliases
Chromosome Location
Chr. 19: 4867614 - 4915469 on Build GRCh38
UniGene ID
Hs.574574
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080523.1 NP_001073992.1 1, 2 302
XM_017027144.1 XP_016882633.1 1, 2 516
XM_017027145.1 XP_016882634.1 1, 2 516

View Full Product Details