Gene Transcripts

Gene Symbol
RBBP4
Entrez Gene ID
5928
Gene Name
RB binding protein 4, chromatin remodeling factor
Gene Aliases
NURF55, RBAP48, lin-53
Chromosome Location
Chr. 1: 32651148 - 32686211 on Build GRCh38
UniGene ID
Hs.16003
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135255.1 NP_001128727.1 4 517
NM_001135256.1 NP_001128728.1 4 420
NM_005610.2 NP_005601.1 4 520
GenBank mRNA AK222779.1 - 4 423
AK299251.1 - 4 420
AK312571.1 - 4 458
BC003092.1 - 4 447 3356558
BC015123.1 - 2 181
BC053904.1 - 4 433 6526200
BC075836.1 - 4 388 6338602
BT007309.1 - 4 361
X71810.1 - 4 434
X74262.1 - 4 445

View Full Product Details