Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001078170.2 | NP_001071638.2 | 20 | 3323 | |
XM_017004845.1 | XP_016860334.1 | 24 | 3774 | ||
XM_017004846.1 | XP_016860335.1 | 20 | 3445 | ||
XM_017004847.1 | XP_016860336.1 | 20 | 3442 | ||
XM_017004848.1 | XP_016860337.1 | 24 | 3774 | ||
XM_017004849.1 | XP_016860338.1 | 24 | 3774 | ||
XM_017004850.1 | XP_016860339.1 | 24 | 3774 | ||
GenBank mRNA | AK300276.1 | - | 1 | 110 | |
AK300352.1 | - | 9 | 1663 | ||
AK302429.1 | - | 20 | 3323 | ||
AY354203.1 | - | 7 | 1299 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024457.3 | NP_001019628.3 | 20 | 3277 | |
XM_011532845.2 | XP_011531147.1 | 21 | 3476 | ||
XM_011532849.2 | XP_011531151.1 | 21 | 3476 | ||
XM_011532850.1 | XP_011531152.1 | 21 | 3476 | ||
XM_011532851.2 | XP_011531153.1 | 21 | 3476 | ||
XM_011532852.2 | XP_011531154.1 | 21 | 3476 | ||
XM_017004103.1 | XP_016859592.1 | 21 | 3476 | ||
XM_017004104.1 | XP_016859593.1 | 20 | 3593 | ||
XM_017004105.1 | XP_016859594.1 | 20 | 3441 | ||
XM_017004106.1 | XP_016859595.1 | 20 | 3560 | ||
XM_017004107.1 | XP_016859596.1 | 21 | 3476 | ||
GenBank mRNA | AK303443.1 | - | 6 | 1193 | |
CR749330.1 | - | 20 | 3277 |