Gene Transcripts
- Gene Symbol
- ANKRD30BL
- Entrez Gene ID
- 554226
- Gene Name
- ankyrin repeat domain 30B-like
- Gene Aliases
- ANKRD30BP3, NCRNA00164
- Chromosome Location
- Chr. 2: 132147591 - 132257969 on Build GRCh38
- UniGene ID
- Hs.380689
- Species
- Homo sapiens
- Gene Symbol
- ANKRD30BP2
- Entrez Gene ID
- 149992
- Gene Name
- ankyrin repeat domain 30B pseudogene 2
- Gene Aliases
- C21orf99, CT85, CTSP-1, CTSP1
- Chromosome Location
- Chr. 21: 13038166 - 13118251 on Build GRCh38
- UniGene ID
- Hs.558645
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026916.1 |
|
6 |
928 |
|
GenBank mRNA |
DQ179138.1 |
- |
1 |
143 |
|
- Gene Symbol
- LOC644669
- Entrez Gene ID
- 644669
- Gene Name
- ankyrin repeat domain 30B pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 18: 15313556 - 15325919 on Build GRCh38
- UniGene ID
- Hs.579474
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027417.1 |
|
5 |
475 |
|
GenBank mRNA |
AK097631.1 |
- |
5 |
475 |
|
View Full Product Details