Gene Transcripts
- Gene Symbol
- TLX1NB
- Entrez Gene ID
- 100038246
- Gene Name
- TLX1 neighbor
- Gene Aliases
- APT-B7, TD1, TDI
- Chromosome Location
- Chr. 10: 101089321 - 101141266 on Build GRCh38
- UniGene ID
- Hs.368784
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130722.1 |
|
3 |
1811 |
|
NR_130723.1 |
|
3 |
1782 |
|
GenBank mRNA |
Y11608.1 |
- |
1 |
336 |
|
View Full Product Details