Gene Transcripts

Gene Symbol
Aebp1
Entrez Gene ID
11568
Gene Name
AE binding protein 1
Gene Aliases
ACLP
Chromosome Location
Chr. 11: 5861853 - 5872225 on Build GRCm38
UniGene ID
Mm.3317
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF053943.1 - 19 2845
AK004712.1 - 15 1854
AK128980.1 - 19 2921
AK158276.1 - 19 2871
AK159330.1 - 19 2852
AK159342.1 - 19 2871
AK159377.1 - 19 2842
AK159409.1 - 19 2872
AK159774.1 - 19 2843
AK159957.1 - 19 2843
AK204340.1 - 2 148
AK210444.1 - 2 149
AK216593.1 - 2 148
BC082577.1 - 19 2857 30648530
X80478.1 - 10 1575

View Full Product Details