Gene Transcripts

Gene Symbol
Aire
Entrez Gene ID
11634
Gene Name
autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
Gene Aliases
Chromosome Location
Chr. 10: 78030022 - 78043610 on Build GRCm38
UniGene ID
Mm.35300
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271549.1 NP_001258478.1 6 793
NM_001271550.1 NP_001258479.1 6 793
NM_001271551.1 NP_001258480.1 6 793
NM_001271552.1 NP_001258481.1 6 793
NM_001271553.1 NP_001258482.1 6 793
NM_001271554.1 NP_001258483.1 6 793
NM_001271555.1 NP_001258484.1 6 793
NM_001271556.1 NP_001258485.1 6 793
NM_001271557.1 NP_001258486.1 6 793
NM_001271558.1 NP_001258487.1 6 793
NM_001271559.1 NP_001258488.1 6 793
NM_009646.2 NP_033776.1 6 793
GenBank mRNA AF079536.1 - 6 763
AF128115.1 - 6 763
AF128116.1 - 6 763
AF128117.1 - 6 763
AF128118.1 - 6 763
AF128119.1 - 6 763
AF128120.1 - 6 763
AF128121.1 - 6 763
AF128122.1 - 6 763
AF128123.1 - 6 763
AF128124.1 - 6 763
AF128125.1 - 6 763
AF128772.1 - 6 706
AF128773.1 - 6 706
AJ132243.1 - 6 706
AJ243821.1 - 6 706
AK153690.1 - 7 847
BC103510.1 - 6 755 40039055
BC103511.1 - 6 755 40039054
BC103518.1 - 6 755 40039058
EU625343.3 - 6 723
JF267399.1 - 6 723

View Full Product Details