Gene Transcripts

Gene Symbol
Btk
Entrez Gene ID
12229
Gene Name
Bruton agammaglobulinemia tyrosine kinase
Gene Aliases
AI528679, xid
Chromosome Location
Chr. X: 134542334 - 134583623 on Build GRCm38
UniGene ID
Mm.4475
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013482.2 NP_038510.2 16 1738
GenBank mRNA BC053392.1 - 16 1674
L08967.1 - 16 1714
L10627.1 - 16 1746
L29788.1 - 16 1710

View Full Product Details