Gene Transcripts
- Gene Symbol
- Cacna2d1
- Entrez Gene ID
- 12293
- Gene Name
- calcium channel, voltage-dependent, alpha2/delta subunit 1
- Gene Aliases
- Ca(v)alpha2delta1, Cacna2, Cchl2a
- Chromosome Location
- Chr. 5: 15934691 - 16374511 on Build GRCm38
- UniGene ID
- Mm.159842
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001110843.1 |
NP_001104313.1
|
1, 2 |
418 |
|
NM_001110844.1 |
NP_001104314.1
|
1, 2 |
418 |
|
NM_001110845.1 |
NP_001104315.1
|
1, 2 |
418 |
|
NM_001110846.1 |
NP_001104316.1
|
1, 2 |
418 |
|
NM_009784.2 |
NP_033914.1
|
1, 2 |
418 |
|
GenBank mRNA |
AK052898.1 |
- |
1, 2 |
198 |
|
AK164580.1 |
- |
1, 2 |
419 |
|
BC115871.1 |
- |
2, 3 |
193 |
40061614 |
U73483.1 |
- |
1, 2 |
325 |
|
U73484.1 |
- |
1, 2 |
325 |
|
U73485.1 |
- |
1, 2 |
325 |
|
U73486.1 |
- |
1, 2 |
325 |
|
U73487.1 |
- |
1, 2 |
325 |
|
View Full Product Details