Gene Transcripts

Gene Symbol
Car4
Entrez Gene ID
12351
Gene Name
carbonic anhydrase 4
Gene Aliases
AW456718, Ca4
Chromosome Location
Chr. 11: 84957754 - 84966054 on Build GRCm38
UniGene ID
Mm.1641
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007607.2 NP_031633.1 4 519
GenBank mRNA AK154754.1 - 4 485
AK165399.1 - 4 447
BC012704.1 - 4 444 4217743

View Full Product Details