Gene Transcripts

Gene Symbol
Lck
Entrez Gene ID
16818
Gene Name
lymphocyte protein tyrosine kinase
Gene Aliases
Hck-3, Lsk, Lskt, p56<lck>, p56Lck
Chromosome Location
Chr. 4: 129548344 - 129573641 on Build GRCm38
UniGene ID
Mm.293753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162432.1 NP_001155904.1 7 683
NM_001162433.1 NP_001155905.1 7 725
NM_010693.3 NP_034823.1 6 780
GenBank mRNA AK088001.1 - 6 781
AK166263.1 - 1 170
BC011474.1 - 7 690 4191610
CA577097.1 - 4 416
M12056.1 - 6 756
X03533.1 - 6 786

View Full Product Details