Gene Transcripts

Gene Symbol
Mybl2
Entrez Gene ID
17865
Gene Name
myeloblastosis oncogene-like 2
Gene Aliases
B-Myb, Bmyb
Chromosome Location
Chr. 2: 163054333 - 163084688 on Build GRCm38
UniGene ID
Mm.4594
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008652.2 NP_032678.1 12 1978
GenBank mRNA AK028497.1 - 12 1978
BC050842.1 - 12 1924 6518298
X70472.1 - 12 1926

View Full Product Details