Gene Transcripts

Gene Symbol
Olfr61
Entrez Gene ID
18362
Gene Name
olfactory receptor 61
Gene Aliases
IH7, MOR253-1
Chromosome Location
Chr. 7: 140637703 - 140638638 on Build GRCm38
UniGene ID
Mm.222845
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146964.1 NP_667175.1 1 65
GenBank mRNA BC119308.1 - 1 161
BC119334.1 - 1 161 8734083

View Full Product Details