Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011516.2 | NP_035646.2 | 13 | 1076 | |
GenBank mRNA | AK015000.1 | - | 13 | 1077 | |
AK133229.1 | - | 13 | 1077 | ||
BC137967.1 | - | 13 | 1038 | 8860986 | |
D88539.1 | - | 8 | 666 | ||
L41069.1 | - | 13 | 1057 | ||
Z38118.1 | - | 13 | 1067 |