Gene Transcripts

Gene Symbol
Wnt1
Entrez Gene ID
22408
Gene Name
wingless-type MMTV integration site family, member 1
Gene Aliases
Int-1, Wnt-1, sw, swaying
Chromosome Location
Chr. 15: 98788074 - 98793837 on Build GRCm38
UniGene ID
Mm.1123
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021279.4 NP_067254.1 4 1224
GenBank mRNA AK141669.1 - 4 1079
BC005449.1 - 4 1213 3493095
M11943.1 - 4 1076

View Full Product Details