Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011794.3 | NP_035924.2 | 6 | 513 | |
GenBank mRNA | AF125043.1 | - | 6 | 471 | |
AK008091.1 | - | 5 | 366 | ||
AK008714.1 | - | 6 | 494 | ||
AK093938.1 | - | 6 | 499 | ||
AK150877.1 | - | 6 | 493 | ||
AK151931.1 | - | 6 | 517 | ||
AK152009.1 | - | 6 | 493 | ||
AK154435.1 | - | 6 | 493 | ||
BC011036.1 | - | 6 | 466 | 4160742 |