Gene Transcripts

Gene Symbol
Azi2
Entrez Gene ID
27215
Gene Name
5-azacytidine induced gene 2
Gene Aliases
AA410145, AZ2
Chromosome Location
Chr. 9: 118040401 - 118069798 on Build GRCm38
UniGene ID
Mm.92705
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048146.2 NP_001041611.1 4 853
NM_001286508.1 NP_001273437.1 4 771
NM_013727.4 NP_038755.1 4 853
GenBank mRNA AB007141.1 - 4 689
AK004992.1 - 4 832
AK015942.1 - 4 674
AK030076.1 - 4 685
AK134714.1 - 4 756
AK153477.1 - 4 817
AK165549.1 - 4 739
AK167783.1 - 4 797
BC006851.1 - 4 748 3156857

View Full Product Details