Gene Transcripts

Gene Symbol
Amotl2
Entrez Gene ID
56332
Gene Name
angiomotin-like 2
Gene Aliases
AW549739, C79691, Lccp, MASCOT, mKIAA0989
Chromosome Location
Chr. 9: 102717294 - 102733418 on Build GRCm38
UniGene ID
Mm.21145
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019764.2 NP_062738.2 4 1287
GenBank mRNA AF175967.1 - 4 570
AF175968.1 - 2 1427
AK040912.1 - 4 1368
AK077535.1 - 4 1283
AK141348.1 - 4 1287
AK164279.1 - 4 1356
AK164819.1 - 4 1287
AK169419.1 - 4 1287
AX763686.1 - 3 1126
BC027824.1 - 4 1257 5323888
BC035201.1 - 4 1269
BC108411.1 - 4 1321

View Full Product Details