Gene Transcripts

Gene Symbol
Asb1
Entrez Gene ID
65247
Gene Name
ankyrin repeat and SOCS box-containing 1
Gene Aliases
1700029O08Rik, 1700054C17Rik, AI426616, AI851378, mKIAA1146
Chromosome Location
Chr. 1: 91539459 - 91559590 on Build GRCm38
UniGene ID
Mm.234152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039126.2 NP_001034215.1 4 777
NM_023046.4 NP_075533.3 4 578
GenBank mRNA AF155352.1 - 4 529
AK006515.1 - 4 579
AK122456.1 - 4 633
AK153774.1 - 4 799
BC034869.1 - 5 572
BC049699.1 - 4 578
BU938650.1 - 4 581
KC789642.1 - 3 307

View Full Product Details