Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025605.3 | NP_079881.2 | 2 | 287 | |
GenBank mRNA | AK003509.1 | - | 2 | 179 | |
AK009320.1 | - | 2 | 176 | ||
AK010247.1 | - | 2 | 220 | ||
AK013389.1 | - | 2 | 197 | ||
AK019947.1 | - | 2 | 138 | ||
AK144476.1 | - | 2 | 186 | ||
AK185971.1 | - | 2 | 130 | ||
AK203171.1 | - | 2 | 129 | ||
BC020142.1 | - | 2 | 193 | 3709051 | |
BC092062.1 | - | 2 | 179 | 4024506 | |
BC096412.1 | - | 2 | 159 | 5255882 | |
BC126973.1 | - | 2 | 194 | 40100355 |