Gene Transcripts

Gene Symbol
Naaa
Entrez Gene ID
67111
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
2210023K21Rik, 3830414F09Rik, Asahl
Chromosome Location
Chr. 5: 92257660 - 92278181 on Build GRCm38
UniGene ID
Mm.28890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163687.1 NP_001157159.1 3 536
NM_025972.4 NP_080248.2 3 542
GenBank mRNA AB162194.1 - 3 508
AK008776.1 - 3 544
AK017141.1 - 3 521
AK133485.1 - 3 527
AK133658.1 - 3 544
AK146191.1 - 3 530

View Full Product Details