Gene Transcripts
- Gene Symbol
- Cap2
- Entrez Gene ID
- 67252
- Gene Name
- CAP, adenylate cyclase-associated protein, 2 (yeast)
- Gene Aliases
- 2810452G09Rik, AV045235
- Chromosome Location
- Chr. 13: 46501903 - 46650281 on Build GRCm38
- UniGene ID
- Mm.44529
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026056.4 |
NP_080332.1
|
12, 13 |
1653 |
|
GenBank mRNA |
AK010235.1 |
- |
12, 13 |
1400 |
|
AK013331.1 |
- |
12, 13 |
1410 |
|
AK014979.1 |
- |
8, 9 |
1048 |
|
AK033071.1 |
- |
12, 13 |
1409 |
|
AK035556.1 |
- |
12, 13 |
1654 |
|
AK047701.1 |
- |
10, 11 |
1220 |
|
AK049813.1 |
- |
9, 10 |
1096 |
|
AK076737.1 |
- |
8, 9 |
1073 |
|
BC050752.1 |
- |
12, 13 |
1414 |
6533812 |
BC057937.1 |
- |
12, 13 |
1468 |
5325599 |
View Full Product Details