Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026395.1 | NP_080671.1 | 6 | 515 | |
GenBank mRNA | AK002550.1 | - | 6 | 515 | |
AK003682.1 | - | 6 | 537 | ||
AK013262.1 | - | 6 | 572 | ||
AK078396.1 | - | 6 | 578 | ||
AK168192.1 | - | 7 | 734 | ||
BC029189.1 | - | 5 | 421 | 4036721 |