Gene Transcripts

Gene Symbol
Angel1
Entrez Gene ID
68737
Gene Name
angel homolog 1 (Drosophila)
Gene Aliases
1110030H02Rik, mKIAA0759
Chromosome Location
Chr. 12: 86700502 - 86734187 on Build GRCm38
UniGene ID
Mm.63979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144524.2 NP_653107.2 3 885
GenBank mRNA AK143549.1 - 3 886
AK220354.1 - 1 16
BC019148.1 - 3 900 5052974

View Full Product Details