Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161329.1 | NP_001154801.1 | 2 | 309 | |
NM_133702.2 | NP_598463.2 | 2 | 309 | ||
GenBank mRNA | AK005123.1 | - | 2 | 199 | |
AK038696.1 | - | 2 | 200 | ||
AK076704.1 | - | 2 | 310 | ||
AK078524.1 | - | 2 | 200 | ||
AK157743.1 | - | 2 | 249 | ||
BC024874.1 | - | 2 | 188 | 3984027 |