Gene Transcripts

Gene Symbol
Prr16
Entrez Gene ID
71373
Gene Name
proline rich 16
Gene Aliases
5430406M13Rik, AI607429
Chromosome Location
Chr. 18: 51117898 - 51305129 on Build GRCm38
UniGene ID
Mm.63546
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081224.2 NP_001074693.1 2 701
GenBank mRNA AK017282.1 - 1 515
AK042678.1 - 863
AK045646.1 - 1 655
AK046456.1 - 2 866
AK081911.1 - 677
BC132481.1 - 2 701 40130758

View Full Product Details