Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001085383.1 | NP_001078852.1 | 11 | 964 | |
NM_023628.2 | NP_076117.2 | 11 | 943 | ||
GenBank mRNA | AJ401160.1 | - | 11 | 897 | |
AK003395.1 | - | 11 | 964 | ||
AK010128.1 | - | 11 | 944 | ||
BC062140.1 | - | 11 | 954 | 30290956 |