Gene Transcripts

Gene Symbol
MYL4
Entrez Gene ID
4635
Gene Name
myosin light chain 4
Gene Aliases
ALC1, AMLC, GT1, PRO1957
Chromosome Location
Chr. 17: 47209017 - 47227650 on Build GRCh38
UniGene ID
Hs.463300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002841.1 NP_001002841.1 3 282
NM_002476.2 NP_002467.1 2 229
XM_005257391.4 XP_005257448.1 3 209
GenBank mRNA AF116676.2 - 581
BC030228.1 - 2 218 5226518
BU658678.1 - 3 288
H83803.1 - 3 282
M36172.1 - 2 210
X13955.1 - 2 210
X52005.1 - 371

View Full Product Details