Gene Transcripts

Gene Symbol
Lin37
Entrez Gene ID
75660
Gene Name
lin-37 homolog (C. elegans)
Gene Aliases
1810054G18Rik
Chromosome Location
Chr. 7: 30555441 - 30559719 on Build GRCm38
UniGene ID
Mm.379242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290569.1 NP_001277498.1 3, 4 461
NM_029377.2 NP_083653.1 3, 4 335
GenBank mRNA AK007858.1 - 3, 4 336
AY428767.1 - 3, 4 185
BC013546.1 - 3, 4 317 4210417
BY707863.1 - 3, 4 335

View Full Product Details