Gene Transcripts

Gene Symbol
Ms4a18
Entrez Gene ID
76002
Gene Name
membrane-spanning 4-domains, subfamily A, member 18
Gene Aliases
5033428B15Rik
Chromosome Location
Chr. 19: 10997024 - 11018026 on Build GRCm38
UniGene ID
Mm.139320
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251849.1 NP_001238778.1 2 220
GenBank mRNA AK017199.1 - 2 222

View Full Product Details