Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030112.2 | NP_084388.2 | 11, 12 | 1493 | |
GenBank mRNA | AK135633.1 | - | 5, 6 | 561 | |
BC117848.1 | - | 11, 12 | 1345 | 40093717 | |
BC117849.1 | - | 11, 12 | 1345 | 40093724 | |
CN839001.1 | - | 3, 4 | 256 |