Gene Transcripts

Gene Symbol
Rab1b
Entrez Gene ID
76308
Gene Name
RAB1B, member RAS oncogene family
Gene Aliases
1110011F09Rik
Chromosome Location
Chr. 19: 5099207 - 5106996 on Build GRCm38
UniGene ID
Mm.182563
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029576.3 NP_083852.1 6 565
GenBank mRNA AB232584.1 - 5 507
AK003609.1 - 6 566
AK042020.1 - 6 567
AK156726.1 - 6 588
BC016408.1 - 6 560 4242206

View Full Product Details