Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133351.3 | NP_579929.1 | 4 | 732 | |
GenBank mRNA | AB038244.1 | - | 4 | 713 | |
AF188613.1 | - | 4 | 646 | ||
AF378085.2 | - | 4 | 723 | ||
AK010640.1 | - | 4 | 615 | ||
AK078696.1 | - | 4 | 687 | ||
AY335911.1 | - | 5 | 580 | ||
BC003851.1 | - | 4 | 706 | 3600399 |