Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278669.1 | NP_001265598.1 | 6 | 2339 | |
NM_001278670.1 | NP_001265599.1 | 6 | 2339 | ||
NM_001278672.1 | NP_001265601.1 | 6 | 1965 | ||
NM_001278673.1 | NP_001265602.1 | 5 | 866 | ||
NM_001278675.1 | NP_001265604.1 | 6 | 1965 | ||
NM_006162.4 | NP_006153.2 | 6 | 2339 | ||
NM_172387.2 | NP_765975.1 | 6 | 1965 | ||
NM_172388.2 | NP_765976.1 | 5 | 1240 | ||
NM_172389.2 | NP_765977.1 | 6 | 1965 | ||
NM_172390.2 | NP_765978.1 | 6 | 2339 | ||
XM_017025783.1 | XP_016881272.1 | 6 | 2181 | ||
GenBank mRNA | AK292641.1 | - | 6 | 2181 | |
AK293760.1 | - | 5 | 811 | ||
BC104753.1 | - | 6 | 1980 | 8143756 | |
BC112243.1 | - | 6 | 1980 | 8327711 | |
BC143457.1 | - | 6 | 1895 | ||
EU887559.1 | - | 6 | 1886 | ||
EU887560.1 | - | 6 | 1847 | ||
EU887561.1 | - | 6 | 1886 | ||
EU887562.1 | - | 6 | 1847 | ||
EU887563.1 | - | 6 | 1886 | ||
EU887564.1 | - | 6 | 1847 | ||
EU887565.1 | - | 6 | 1886 | ||
EU887566.1 | - | 6 | 1847 | ||
U08015.1 | - | 6 | 2125 | ||
U59736.1 | - | 6 | 1922 | ||
U80917.1 | - | 6 | 2255 | ||
U80918.1 | - | 6 | 1909 | ||
U80919.1 | - | 5 | 1068 |