Gene Transcripts

Gene Symbol
Cacng8
Entrez Gene ID
81905
Gene Name
calcium channel, voltage-dependent, gamma subunit 8
Gene Aliases
mir-935
Chromosome Location
Chr. 7: 3394117 - 3415605 on Build GRCm38
UniGene ID
Mm.357990
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133190.1 NP_573453.1 2 409
GenBank mRNA AF361350.1 - 2 409

View Full Product Details