Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029612.4 | NP_083888.3 | 3 | 578 | |
GenBank mRNA | AB196824.1 | - | 3 | 441 | |
AB196825.1 | - | 3 | 441 | ||
AB196826.1 | - | 3 | 441 | ||
AB196827.1 | - | 3 | 441 | ||
AK009505.1 | - | 3 | 534 | ||
AK090041.1 | - | 3 | 569 | ||
BC019477.1 | - | 3 | 517 | 4458767 |