Gene Transcripts

Gene Symbol
Rell1
Entrez Gene ID
100532
Gene Name
RELT-like 1
Gene Aliases
AA536743
Chromosome Location
Chr. 5: 63888667 - 63968900 on Build GRCm38
UniGene ID
Mm.243632
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145923.4 NP_666035.2 3 540
GenBank mRNA AK133845.1 - 4 560
AK140108.1 - 3 540
AK159944.1 - 3 466
AK161061.1 - 3 466
AK182372.1 - 2 99
AK202598.1 - 2 99
BC031198.1 - 3 509 5059693
BC066137.1 - 3 519 30012041
BC066160.1 - 3 519 30012671

View Full Product Details