Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004302.4 | NP_004293.1 | 4 | 824 | |
NM_020327.3 | NP_064732.3 | 4 | 776 | ||
NM_020328.3 | NP_064733.3 | 4 | 824 | ||
XM_011538966.2 | XP_011537268.1 | 4 | 802 | ||
XM_011538967.2 | XP_011537269.1 | 4 | 802 | ||
XM_017020199.1 | XP_016875688.1 | 4 | 1522 | ||
XM_017020200.1 | XP_016875689.1 | 5 | 917 | ||
XM_017020201.1 | XP_016875690.1 | 4 | 802 | ||
GenBank mRNA | AK299120.1 | - | 4 | 787 | |
AK299496.1 | - | 4 | 776 | ||
BC000254.1 | - | 4 | 773 | 3352720 | |
BC040531.1 | - | 4 | 821 | 5260767 | |
BT007072.1 | - | 4 | 747 | ||
L10125.1 | - | 4 | 786 | ||
L10126.1 | - | 4 | 747 | ||
U14722.1 | - | 4 | 747 | ||
Z22536.1 | - | 4 | 747 |